EMAIL THIS PAGE TO A FRIEND
72066 Sigma-Aldrich

Neoabietic acid

analytical standard

  • CAS Number 471-77-2

  • Empirical Formula (Hill Notation) C20H30O2

  • Molecular Weight 302.45

  •  Beilstein Registry Number 2702391

  •  MDL number MFCD01699019

  •  PubChem Substance ID 329763737

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Biochemicals and Reagents, Fatty Acids, Isoprenoids,
grade   analytical standard
InChI Key   KGMSWPSAVZAMKR-RMGPQRKISA-N
assay   ≥98.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. 72066.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TP8740000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?