• USA Home
  • 72247 - Nickel(II) chloride hexahydrate

EMAIL THIS PAGE TO A FRIEND
72247 Sigma-Aldrich

Nickel(II) chloride hexahydrate

purum p.a., ≥98.0% (KT)

Purchase

Properties

Related Categories Essential Chemicals, Nickel Salts, Research Essentials, Salts
InChI Key   LAIZPRYFQUWUBN-UHFFFAOYSA-L
grade   purum p.a.
assay   ≥98.0% (KT)
form   powder or crystals
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤500 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN3288 - class 6.1 - PG 3 - EHS - RQ - Toxic solid, inorganic, n.o.s., HI: all
WGK Germany 
3
RTECS 
QR6480000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?