• USA Home
  • 72280 - Nickel(II) sulfate hexahydrate

EMAIL THIS PAGE TO A FRIEND
72280 Sigma-Aldrich

Nickel(II) sulfate hexahydrate

purum p.a., crystallized, ≥99.0% (KT)

Purchase

Properties

Related Categories Essential Chemicals, Nickel Salts, Research Essentials, Salts
vapor density   2.07 (vs air)
InChI Key   RRIWRJBSCGCBID-UHFFFAOYSA-L
grade   purum p.a.
assay   ≥99.0% (KT)
quality   crystallized
anion traces   chloride (Cl-): ≤100 mg/kg
cation traces   Ca: ≤50 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Na: ≤1000 mg/kg
  Pb: ≤50 mg/kg
  Zn: ≤50 mg/kg

Price and Availability


Laboratory Gloves

Redi-Dri
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?