EMAIL THIS PAGE TO A FRIEND
72310 Sigma-Aldrich

Nicotinic acid

purum, ≥99.0% (HPLC)

Synonym: 3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

  • CAS Number 59-67-6

  • Empirical Formula (Hill Notation) C6H5NO2

  • Molecular Weight 123.11

  •  Beilstein Registry Number 109591

  •  EC Number 200-441-0

  •  MDL number MFCD00006391

  •  PubChem Substance ID 329763975

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C6, Chemical Synthesis, Heterocyclic Building Blocks,
InChI Key   PVNIIMVLHYAWGP-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (HPLC)
mp   233-238 °C
  236-239 °C(lit.)

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
QT0525000
Flash Point(F) 
379.4 °F
Flash Point(C) 
193 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?