EMAIL THIS PAGE TO A FRIEND
72347 Sigma-Aldrich

Nicotinamide

tested according to Ph.Eur.

Synonym: Niacinamide, Nicotinamidum, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP

  • CAS Number 98-92-0

  • Empirical Formula (Hill Notation) C6H6N2O

  • Molecular Weight 122.12

  •  Beilstein Registry Number 383619

  •  EC Number 202-713-4

  •  MDL number MFCD00006395

  •  PubChem Substance ID 329764011

Purchase

Properties

Related Categories Biochemicals and Reagents, Core Bioreagents, Research Essentials, Vitamin B, Vitamin B3,
InChI Key   DFPAKSUCGFBDDF-UHFFFAOYSA-N
assay   ≥99%
form   crystalline
pharmacopeia   testing conforms to Pharmacopeia
mp   128-131 °C(lit.)
Gene Information   human ... PARP1(142), SIRT2(22933)

Description

Biochem/physiol Actions

Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
QS3675000
Flash Point(F) 
302 °F
Flash Point(C) 
150 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?