EMAIL THIS PAGE TO A FRIEND
72385 Sigma-Aldrich

Calcium ionophore VI

Selectophore, function tested

Synonym: tert-Butyl-calix[4]arene tetrakis[2-(diphenylphosphoryl)ethyl ether]

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Ionophores Potentiometric for ISE More...
grade   Selectophore
InChI Key   JTLQADIQVNKIFC-UHFFFAOYSA-N

Description

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

21059 Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?