• USA Home
  • 72470 - Nigrosin water soluble

EMAIL THIS PAGE TO A FRIEND
72470 Sigma-Aldrich

Nigrosin water soluble

for microscopy (Bact., Hist.)

Synonym: Acid black 2

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, N, Stains & Dyes, A to Z, Stains and Dyes More...
grade   for microscopy (Bact., Hist.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
GC4762250

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?