EMAIL THIS PAGE TO A FRIEND
72482 Sigma-Aldrich

Nile Blue chloride

for fluorescence

  • CAS Number 2381-85-3

  • Empirical Formula (Hill Notation) C20H20ClN3O

  • Molecular Weight 353.85

  •  Beilstein Registry Number 3923952

  •  MDL number MFCD00011936

  •  PubChem Substance ID 329764109

Purchase

Properties

Related Categories Biochemicals and Reagents, Luminescent Compounds/Detection, Stains and Dyes More...
grade   for fluorescence
InChI Key   SHXOKQKTZJXHHR-UHFFFAOYSA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DJ1930000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

222550 Nile Blue chloride, Dye content 85 %

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?