EMAIL THIS PAGE TO A FRIEND
72490 Sigma-Aldrich

Ninhydrin

ACS reagent, ≥98.0% (UV)

Synonym: 1,2,3-Indantrione monohydrate, 2,2-Dihydroxy-1,3-indanedione, Trioxohydrindene monohydrate

  • CAS Number 485-47-2

  • Empirical Formula (Hill Notation) C9H6O4

  • Molecular Weight 178.14

  •  Beilstein Registry Number 1910963

  •  EC Number 207-618-1

  •  MDL number MFCD00003791

  •  PubChem Substance ID 329764112

Purchase

Description

Application

Used for the detection of free amino groups in amino acids, peptides and proteins.

suitable for Kaiser test

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NK5425000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?