EMAIL THIS PAGE TO A FRIEND
72537 Sigma-Aldrich

2-Methoxypropene

analytical standard

Synonym: Isopropenyl methyl ether

  • CAS Number 116-11-0

  • Linear Formula CH2=C(CH3)OCH3

  • Molecular Weight 72.11

  •  Beilstein Registry Number 1734635

  •  EC Number 204-125-3

  •  MDL number MFCD00014929

  •  PubChem Substance ID 24886415

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chemical Synthesis,
grade   analytical standard
vapor pressure   27.96 psi ( 55 °C)
  8.28 psi ( 20 °C)
InChI Key   YOWQWFMSQCOSBA-UHFFFAOYSA-N
assay   ≥99.5% (GC)
form   neat
autoignition temp.   374 °F
shelf life   limited shelf life, expiry date on the label
contains   potassium carbonate as stabilizer
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.382(lit.)
  n20/D 1.382
bp   34-36 °C(lit.)
density   0.753 g/mL at 25 °C(lit.)
format   neat
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Nylon Membrane and Net Filters
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 1993A 3 / PGI
WGK Germany 
1
RTECS 
UD0800000
Flash Point(F) 
-20.2 °F
Flash Point(C) 
-29 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?