EMAIL THIS PAGE TO A FRIEND
72632 Sigma-Aldrich

2-Nitroethanol

purum, ≥95.0% (GC)

  • CAS Number 625-48-9

  • Linear Formula NO2CH2CH2OH

  • Molecular Weight 91.07

  •  Beilstein Registry Number 1633753

  •  EC Number 210-895-1

  •  MDL number MFCD00007405

  •  PubChem Substance ID 24886434

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
InChI Key   KIPMDPDAFINLIV-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.445(lit.)
  n20/D 1.445
bp   194 °C/765 mmHg(lit.)
mp   −80 °C(lit.)
density   1.27 g/mL at 25 °C(lit.)

Description

Other Notes

Preparation of nitroethylene, a versatile reactive reagent, fairly stable in solution

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
KL9370000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

146633 2-Nitroethanol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?