• USA Home
  • 72640 - 1-Ethyl-2-nitrobenzene

EMAIL THIS PAGE TO A FRIEND
72640 Sigma-Aldrich

1-Ethyl-2-nitrobenzene

purum, ≥98.0% (GC)

Synonym: 2-Nitroethylbenzene

  • CAS Number 612-22-6

  • Linear Formula C2H5C6H4NO2

  • Molecular Weight 151.16

  •  Beilstein Registry Number 1865655

  •  EC Number 210-299-1

  •  MDL number MFCD00024294

  •  PubChem Substance ID 24886435

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitro Compounds, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   PXWYZLWEKCMTEZ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.536
  n20/D 1.537(lit.)
bp   172-174 °C/18 mmHg(lit.)
mp   -13--10 °C(lit.)
density   1.127 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
228.2 °F
Flash Point(C) 
109 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?