EMAIL THIS PAGE TO A FRIEND
72750 Sigma-Aldrich

Nω-Nitro-L-arginine

≥99.0% (NT)

Synonym: L-NNA, N5-(Nitroamidino)-L-2,5-diaminopentanoic acid, NG-NO2-L-Arg

  • CAS Number 2149-70-4

  • Empirical Formula (Hill Notation) C6H13N5O4

  • Molecular Weight 219.20

  •  Beilstein Registry Number 1728914

  •  EC Number 218-418-9

  •  MDL number MFCD00007033

  •  PubChem Substance ID 329764324

Purchase

Properties

Related Categories Amino Acids & Derivatives, Arginine, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   MRAUNPAHJZDYCK-BYPYZUCNSA-N
assay   ≥99.0% (NT)
optical activity   [α]20/D +24±1°, c = 4% in 2 M HCl
ign. residue   ≤0.05%
mp   255-260 °C

Description

Biochem/physiol Actions

Irreversible inhibitor of constitutive nitric oxide synthase (nNOS) and a reversible inhibitor of inducible nitric oxide synthase (iNOS).

Other Notes

Nitric oxide synthase inhibitor

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?