EMAIL THIS PAGE TO A FRIEND
72790 Sigma-Aldrich

3-Nitrobenzaldehyde

technical, ≥95% (GC)

  • CAS Number 99-61-6

  • Linear Formula O2NC6H4CHO

  • Molecular Weight 151.12

  •  Beilstein Registry Number 386795

  •  EC Number 202-772-6

  •  MDL number MFCD00007249

  •  PubChem Substance ID 329764361

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C7, Carbonyl Compounds, Chemical Synthesis,
InChI Key   ZETIVVHRRQLWFW-UHFFFAOYSA-N
grade   technical
assay   ≥95% (GC)
impurities   1% 3-nitrobenzoic acid
mp   55-58 °C(lit.)
  55-58 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
CU7250000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?