EMAIL THIS PAGE TO A FRIEND
72802 Sigma-Aldrich

4-Nitrobenzaldehyde

purum, ≥97.0% (HPLC)

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C7, Carbonyl Compounds, Chemical Synthesis,
InChI Key   BXRFQSNOROATLV-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
mp   103-106 °C(lit.)
  103-106 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
CU7350000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?