EMAIL THIS PAGE TO A FRIEND
72895 Sigma-Aldrich

2-Nitrobenzoic acid

technical, ≥85% (HPLC)

  • CAS Number 552-16-9

  • Linear Formula O2NC6H4CO2H

  • Molecular Weight 167.12

  •  Beilstein Registry Number 1910632

  •  EC Number 209-004-9

  •  MDL number MFCD00007137

  •  PubChem Substance ID 24886461

Purchase

Properties

Related Categories Building Blocks, C7, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   SLAMLWHELXOEJZ-UHFFFAOYSA-N
grade   technical
assay   ≥85% (HPLC)
mp   146-148 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DH5050000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?