EMAIL THIS PAGE TO A FRIEND
72902 Sigma-Aldrich

3-Nitrobenzoic acid

purum, ≥98.0% (HPLC)

  • CAS Number 121-92-6

  • Linear Formula O2NC6H4CO2H

  • Molecular Weight 167.12

  •  Beilstein Registry Number 908644

  •  EC Number 204-508-5

  •  MDL number MFCD00007251

Purchase

Properties

Related Categories Building Blocks, C7, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   AFPHTEQTJZKQAQ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   139-141 °C(lit.)
  141-144 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DH5000000
Flash Point(F) 
374 °F
Flash Point(C) 
190 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

185329 3-Nitrobenzoic acid, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?