• USA Home
  • 73030 - 2-Nitrobenzenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
73030 Sigma-Aldrich

2-Nitrobenzenesulfonyl chloride

purum, ≥98.0% (AT)

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonyl Halides, Sulfur Compounds More...
InChI Key   WPHUUIODWRNJLO-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
mp   63-67 °C(lit.)
  65-67 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

N11507 2-Nitrobenzenesulfonyl chloride, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?