EMAIL THIS PAGE TO A FRIEND
73080 Sigma-Aldrich

4-Nitrobenzonitrile

purum, ≥98.0% (GC)

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   NKJIFDNZPGLLSH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
mp   144-147 °C(lit.)
  146-150 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3439 6.1 / PGII
WGK Germany 
3
RTECS 
DI4903500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N12007 4-Nitrobenzonitrile, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?