• USA Home
  • 73110 - 3-Nitrobenzoyl chloride

EMAIL THIS PAGE TO A FRIEND
73110 Sigma-Aldrich

3-Nitrobenzoyl chloride

purum, ≥99.0% (T)

  • CAS Number 121-90-4

  • Linear Formula O2NC6H4COCl

  • Molecular Weight 185.56

  •  Beilstein Registry Number 777186

  •  EC Number 204-505-9

  •  MDL number MFCD00007247

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   NXTNASSYJUXJDV-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
bp   275-278 °C(lit.)
mp   31-34 °C(lit.)
  31-34 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Risk of explosion if heated under confinement.
RIDADR 
UN 2923 6.1(8) / PGIII
WGK Germany 
3
RTECS 
DM6646000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

127663 3-Nitrobenzoyl chloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?