• USA Home
  • 73170 - 4-Nitrobenzyl bromide

EMAIL THIS PAGE TO A FRIEND
73170 Sigma-Aldrich

4-Nitrobenzyl bromide

purum, ≥97.0% (HPLC)

Synonym: α-Bromo-4-nitrotoluene

  • CAS Number 100-11-8

  • Linear Formula O2NC6H4CH2Br

  • Molecular Weight 216.03

  •  Beilstein Registry Number 742796

  •  EC Number 202-820-6

  •  MDL number MFCD00007373

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitro Compounds, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   VOLRSQPSJGXRNJ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
mp   96-99 °C(lit.)
  96-99 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
RTECS 
XS7967000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

N13054 4-Nitrobenzyl bromide, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?