• USA Home
  • 73210 - 4-(4-Nitrobenzyl)pyridine

EMAIL THIS PAGE TO A FRIEND
73210 Sigma-Aldrich

4-(4-Nitrobenzyl)pyridine

for TLC derivatization, for spectrophotometric det. of phosphorus-containing pesticides

  • CAS Number 1083-48-3

  • Empirical Formula (Hill Notation) C12H10N2O2

  • Molecular Weight 214.22

  •  Beilstein Registry Number 170877

  •  EC Number 214-108-2

  •  MDL number MFCD00006445

  •  PubChem Substance ID 24886523

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents TLC, Spectroscopy,
grade   for TLC derivatization
InChI Key   MNHKUCBXXMFQDM-UHFFFAOYSA-N
assay   ≥98.0% (NT)
quality   for spectrophotometric det. of phosphorus-containing pesticides
ign. residue   ≤0.1%
mp   69-71 °C(lit.)

Description

Application

for the detection of epoxides and methylating reagents; for spectrophotometric determination of phosphorus-containing pesticides

Other Notes

Reagent for the determination of alkylating agents

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Just Prove it! Spectroquant Prove
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UT6360000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?