EMAIL THIS PAGE TO A FRIEND
73334 Sigma-Aldrich

Imbentin-N/63

in accordance for TLC

Synonym: 4-Nonylphenyl-polyethylene glycol, Tergitol NPX replacement product

Purchase

Properties

Related Categories Biochemicals and Reagents, Cleaners, Detergents, Detergents A to M, Labware,
description   non-ionic
mol wt   average mol wt 680
refractive index   n20/D 1.49
CMC   0.059 mM(20-25°C)
transition temp   cloud point 45-50 °C
anion traces   chloride (Cl-): ≤500 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤500 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg
suitability   in accordance for TLC

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Legal Information

Tergitol is a trademark of Dow Chemical Company

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
AX0247000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?