EMAIL THIS PAGE TO A FRIEND
73360 Sigma-Aldrich

1-Nitrohexane

purum, ≥98.0% (GC)

  • CAS Number 646-14-0

  • Linear Formula CH3(CH2)5NO2

  • Molecular Weight 131.17

  •  Beilstein Registry Number 1749466

  •  EC Number 211-467-7

  •  MDL number MFCD00007411

  •  PubChem Substance ID 329764830

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitro Compounds, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   FEYJIFXFOHFGCC-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.423(lit.)
  n20/D 1.424
bp   91-92 °C/24 mmHg(lit.)
density   0.94 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
163.4 °F
Flash Point(C) 
73 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

259497 1-Nitrohexane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?