EMAIL THIS PAGE TO A FRIEND
73415 Sigma-Aldrich

5-Nitroindole

purum, ≥97.0% (TLC)

Synonym: NSC 520594

  • CAS Number 6146-52-7

  • Empirical Formula (Hill Notation) C8H6N2O2

  • Molecular Weight 162.15

  •  Beilstein Registry Number 383777

  •  EC Number 228-153-0

  •  MDL number MFCD00005673

  •  PubChem Substance ID 329764876

Purchase

Properties

Related Categories Building Blocks, C7 to C9, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   OZFPSOBLQZPIAV-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (TLC)
mp   140-142 °C(lit.)
  140-142 °C

Description

Application

Reactant for preparation of:
• Pharmaceutically active 2-oxo-1-pyrrolidine analogues
• Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
• Protein Kinase Inhibitors and antiproliferative agents
• Positive Allosteric Modulators of Metabotropic Glutamate Receptor 4 (mGlu4)
• Antifungal agents
• Cannabinoid receptor type 1 (CB1) antagonists
• Potential anticancer agents
• Potential antivascular agents
• Selective Anti-leukemic agents
• Anti human immunodeficiency virus subtype 1 (HIV-1) agents

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NM1168200

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N17602 5-Nitroindole, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?