EMAIL THIS PAGE TO A FRIEND
73500 Sigma-Aldrich

1-Nitronaphthalene

purum, ≥98.0% (GC)

  • CAS Number 86-57-7

  • Empirical Formula (Hill Notation) C10H7NO2

  • Molecular Weight 173.17

  •  Beilstein Registry Number 1867714

  •  EC Number 201-684-5

  •  MDL number MFCD00003913

  •  PubChem Substance ID 329763785

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitro Compounds, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   RJKGJBPXVHTNJL-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   304 °C(lit.)
mp   53-57 °C(lit.)
  56-58 °C
density   1.223 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2538 4.1 / PGIII
WGK Germany 
2
RTECS 
QJ9720000
Flash Point(F) 
327.2 °F
Flash Point(C) 
164 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?