EMAIL THIS PAGE TO A FRIEND
73542 Sigma-Aldrich

2-Nitrophenol

purum, ≥98.0% (HPLC)

  • CAS Number 88-75-5

  • Linear Formula O2NC6H4OH

  • Molecular Weight 139.11

  •  Beilstein Registry Number 775403

  •  EC Number 201-857-5

  •  MDL number MFCD00011688

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
vapor pressure   1 mmHg ( 49.3 °C)
InChI Key   IQUPABOKLQSFBK-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   43-45 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 1663 6.1 / PGIII
WGK Germany 
2
RTECS 
SM2100000
Flash Point(F) 
215.6 °F
Flash Point(C) 
102 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?