EMAIL THIS PAGE TO A FRIEND
73552 Sigma-Aldrich

3-Nitrophenol

purum, ≥98.0% (HPLC)

  • CAS Number 554-84-7

  • Linear Formula O2NC6H4OH

  • Molecular Weight 139.11

  •  Beilstein Registry Number 1907946

  •  EC Number 209-073-5

  •  MDL number MFCD00007240

  •  PubChem Substance ID 24886583

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   RTZZCYNQPHTPPL-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
bp   194 °C/70 mmHg(lit.)
mp   91-97 °C
  96-98 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 1663 6.1 / PGIII
WGK Germany 
3
RTECS 
SM1925000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

163031 3-Nitrophenol, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?