• USA Home
  • 73613 - (S)-α-Methyl-4-nitrobenzylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
73613 Sigma-Aldrich

(S)-α-Methyl-4-nitrobenzylamine hydrochloride

puriss., ≥99.0% (HPLC)

Synonym: (S)-1-(4-Nitrophenyl)ethylamine hydrochloride

Purchase

Properties

Related Categories Amine Salts, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   CZQQGVFHLSBEDV-RGMNGODLSA-N
grade   puriss.
assay   ≥99.0% (HPLC)
optical activity   [α]20/D −7.2±0.5°, c = 1% in 0.05 M NaOH
mp   248-250 °C(lit.)
  248-250 °C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

339822 (S)-α-Methyl-4-nitrobenzylamine hydrochloride, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?