• USA Home
  • 73620 - 2-Nitro-1,4-phenylenediamine

EMAIL THIS PAGE TO A FRIEND
73620 Sigma-Aldrich

2-Nitro-1,4-phenylenediamine

technical, ≥95.0% (NT)

Synonym: 1,4-Diamino-2-nitrobenzene, 2-Nitro-p-phenylenediamine

  • CAS Number 5307-14-2

  • Linear Formula O2NC6H3(NH2)2

  • Molecular Weight 153.14

  •  Beilstein Registry Number 2210195

  •  EC Number 226-164-5

  •  MDL number MFCD00007903

  •  PubChem Substance ID 24886594

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks, Polyamines More...
InChI Key   HVHNMNGARPCGGD-UHFFFAOYSA-N
grade   technical
assay   ≥95.0% (NT)
mp   135-138 °C(lit.)
  135-138 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
ST3000000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

N21200 2-Nitro-1,4-phenylenediamine, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?