• USA Home
  • 73780 - 3-Nitrophthalic anhydride

EMAIL THIS PAGE TO A FRIEND
73780 Sigma-Aldrich

3-Nitrophthalic anhydride

purum, ≥96.0% (NT)

  • CAS Number 641-70-3

  • Empirical Formula (Hill Notation) C8H3NO5

  • Molecular Weight 193.11

  •  Beilstein Registry Number 179963

  •  EC Number 211-373-6

  •  MDL number MFCD00005921

  •  PubChem Substance ID 329765229

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Anhydrides, Chemical Synthesis, Organic Building Blocks More...
InChI Key   ROFZMKDROVBLNY-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (NT)
mp   162-165 °C
  163-165 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?