EMAIL THIS PAGE TO A FRIEND
73805 Sigma-Aldrich

2-Nitro-1-propanol

purum, ≥97.0% (GC)

  • CAS Number 2902-96-7

  • Linear Formula CH3CH(NO2)CH2OH

  • Molecular Weight 105.09

  •  Beilstein Registry Number 1748592

  •  EC Number 220-797-0

  •  MDL number MFCD00007398

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
InChI Key   PCNWBUOSTLGPMI-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
form   liquid (clear)
color   clear green-yellow
refractive index   n20/D 1.439(lit.)
  n20/D 1.440
bp   72-74 °C/1 mmHg(lit.)
density   1.185 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Preparation of 2-nitropropene

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
213.8 °F
Flash Point(C) 
101 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?