• USA Home
  • 74030 - Nitrotetrazolium Blue chloride

EMAIL THIS PAGE TO A FRIEND
74030 Sigma-Aldrich

Nitrotetrazolium Blue chloride

≥88% (T)

Synonym: 2,2′-bis(4-Nitrophenyl)-5,5′-diphenyl-3,3′-(3,3′-dimethoxy-4,4′-diphenylene)ditetrazolium chloride, 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride], p-Nitro-Blue tetrazolium chloride, p-Nitrotetrazolium blue, NBT, Nitro BT

  • CAS Number 298-83-9

  • Linear Formula C40H30N10O6 · 2Cl

  • Molecular Weight 817.64

  •  Beilstein Registry Number 4115923

  •  EC Number 206-067-4

  •  MDL number MFCD00012159

  •  PubChem Substance ID 329765398

Purchase

Description

Other Notes

Enzyme substrate e.g. for the determination of glucose-6-phosphate dehydrogenase; Precipitating agent for fibrinogen

Price and Availability


Laboratory Gloves

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XF8045000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

N6876 Nitrotetrazolium Blue chloride, ~98% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?