• USA Home
  • 74040 - 2-Nitro-5-thiocyanatobenzoic acid

EMAIL THIS PAGE TO A FRIEND
74040 Sigma-Aldrich

2-Nitro-5-thiocyanatobenzoic acid

≥97.0% (T)

Synonym: 2-Nitro-5-thiocyanobenzoic acid, NTCB

  • CAS Number 30211-77-9

  • Linear Formula O2NC6H3(SCN)CO2H

  • Molecular Weight 224.19

  •  Beilstein Registry Number 2124001

  •  EC Number 250-093-9

  •  MDL number MFCD00007139

Purchase

Properties

InChI Key   NQUNIMFHIWQQGJ-UHFFFAOYSA-N
assay   ≥97.0% (T)
mp   156-157 °C(lit.)
  159-163 °C

Description

Other Notes

Reagent for the selective cyanylation of sulfhydryl groups ; Modifies and cleaves peptides at cysteine residues

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

N7009 2-Nitro-5-thiocyanatobenzoic acid, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?