EMAIL THIS PAGE TO A FRIEND
74080 Sigma-Aldrich

4-Nitrotoluene

purum, ≥98.0% (GC)

Synonym: 1-Methyl-4-nitrobenzene

  • CAS Number 99-99-0

  • Linear Formula CH3C6H4NO2

  • Molecular Weight 137.14

  •  Beilstein Registry Number 1906911

  •  EC Number 202-808-0

  •  MDL number MFCD00007366

  •  PubChem Substance ID 329765440

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Nitro Compounds, Nitrogen Compounds, Organic Building Blocks More...
vapor density   4.7 (vs air)
vapor pressure   5 mmHg ( 85 °C)
InChI Key   ZPTVNYMJQHSSEA-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   734 °F
bp   238 °C(lit.)
mp   51-53 °C
  52-54 °C(lit.)
density   1.392 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3446 6.1 / PGII
WGK Germany 
3
RTECS 
XT3325000
Flash Point(F) 
222.8 °F
Flash Point(C) 
106 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N27322 4-Nitrotoluene, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?