• USA Home
  • 74153 - (−)-Noe’s reagent

EMAIL THIS PAGE TO A FRIEND
74153 Sigma-Aldrich

(−)-Noe’s reagent

for chiral derivatization

Synonym: (−)-MBF-OH dimer, (2S,3aR,eS,7aR)-Octahydro-7,8,8-trimethyl-4,7-methanobenzofuran-2-yl ether, Bis[(2S,3aR,4S,7aR)octahydro-7,8,8-trimethyl-4,7-methanobenzofuran-2-yl] ether

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents HPLC, UV-VIS More...
grade   for chiral derivatization
InChI Key   VUDXCBLBKXFCNA-FEFNCVQLSA-N
optical activity   [α]20/D −206±2°, c = 2.5% in THF
mp   ~150 °C
storage temp.   2-8°C

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?