EMAIL THIS PAGE TO A FRIEND
74210 Sigma-Aldrich

Methyl nonadecanoate

puriss., ≥98.5% (GC)

Synonym: Nonadecanoic acid methyl ester

  • CAS Number 1731-94-8

  • Linear Formula CH3(CH2)17COOCH3

  • Molecular Weight 312.53

  •  Beilstein Registry Number 1788184

  •  EC Number 217-056-9

  •  MDL number MFCD00009011

  •  PubChem Substance ID 329765559

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis,
InChI Key   BDXAHSJUDUZLDU-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (GC)
mp   37-40 °C(lit.)
  37-41 °C

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SPB®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SPB®-1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N5377 Methyl nonadecanoate, ≥98% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?