• USA Home
  • 74302 - Poly-(vinyl chloride-co-vinyl acetate-co-hydroxypropyl acrylate)

EMAIL THIS PAGE TO A FRIEND
74302 Sigma-Aldrich

Poly-(vinyl chloride-co-vinyl acetate-co-hydroxypropyl acrylate)

Selectophore, 80% vinyl chloride, 15% hydroxypropyl acrylate and 5% vinyl acetate

Synonym: Poly(vinyl chloride-co-hydroxypropyl acrylate-co-vinyl acetate)

  • CAS Number 39317-41-4

  • Linear Formula [CH2CH(Cl)]x[CH2CH(O2CCH3)]y[CH2CH[CO2CH2CH(OH)CH3]]z

  •  MDL number MFCD03458150

Purchase

Properties

Related Categories Analytical/Chromatography, Ion Sensor Materials, Polymers for ISE More...
grade   Selectophore
mol wt   Mw ~40,000
solubility   MEK: soluble
  THF: soluble
  acetone: soluble
  toluene: soluble

Description

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Polymer membrane material in solid state polyion electrochemical sensors (e.g. heparin, protamine)

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?