EMAIL THIS PAGE TO A FRIEND
74308 Sigma-Aldrich

5-Nonanol

puriss., ≥99.5% (GC)

Synonym: Dibutyl carbinol

  • CAS Number 623-93-8

  • Linear Formula (CH3CH2CH2CH2)2CHOH

  • Molecular Weight 144.25

  •  Beilstein Registry Number 1733552

  •  EC Number 210-820-2

  •  MDL number MFCD00021944

  •  PubChem Substance ID 24886677

Purchase

Properties

Related Categories Alcohols, Building Blocks, C9 to C10, Chemical Synthesis, Organic Building Blocks,
InChI Key   FCBBRODPXVPZAH-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.5% (GC)
refractive index   n20/D 1.429
bp   195 °C(lit.)
density   0.821 g/mL at 20 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
170.6 °F
Flash Point(C) 
77 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?