• USA Home
  • 74381 - (2-Nonen-1-yl)succinic anhydride

EMAIL THIS PAGE TO A FRIEND
74381 Sigma-Aldrich

(2-Nonen-1-yl)succinic anhydride

technical, ≥90% (NT)

Purchase

Properties

Related Categories Building Blocks, C13, C13 to C14, Carbonyl Compounds, Chemical Synthesis,
InChI Key   CIMXGTDNUHWJCZ-BQYQJAHWSA-N
grade   technical
assay   ≥90% (NT)
refractive index   n20/D 1.476
density   1.03 g/mL at 20 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
294.8 °F
Flash Point(C) 
146 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N7644 (2-Nonen-1-yl)succinic anhydride, ≥85% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?