EMAIL THIS PAGE TO A FRIEND
74400 Sigma-Aldrich

Nonylamine

purum, ≥97.0% (GC)

Synonym: 1-Aminononane

  • CAS Number 112-20-9

  • Linear Formula CH3(CH2)8NH2

  • Molecular Weight 143.27

  •  Beilstein Registry Number 1733633

  •  EC Number 203-945-9

  •  MDL number MFCD00008249

  •  PubChem Substance ID 329765669

Purchase

Properties

InChI Key   FJDUDHYHRVPMJZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.433(lit.)
  n20/D 1.434
bp   201 °C(lit.)
density   0.782 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2735 - class 8 - PG 2 - EHS - Amines, liquid, corrosive, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
165.2 °F
Flash Point(C) 
74 °C
Protocols & Articles

Articles

GC Analysis of Primary Amines on Carbowax® Amine

From our library of Articles, Sigma-Aldrich presents GC Analysis of Primary Amines on Carbowax® Amine
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?