EMAIL THIS PAGE TO A FRIEND
74700 Sigma-Aldrich

Octacosanoic acid

puriss., synthetic, ≥98.5% (GC)

Synonym: Montanic acid

  • CAS Number 506-48-9

  • Linear Formula CH3(CH2)26CO2H

  • Molecular Weight 424.74

  •  Beilstein Registry Number 1801616

  •  EC Number 208-041-8

  •  MDL number MFCD00002812

  •  PubChem Substance ID 329765806

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C13 to C42+, Carbonyl Compounds, Carboxylic Acids,
InChI Key   UTOPWMOLSKOLTQ-UHFFFAOYSA-N
grade   puriss.
assay   ≥98.5% (GC)
quality   synthetic
mp   91-93 °C(lit.)
  91-93 °C

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?