EMAIL THIS PAGE TO A FRIEND
74710 Sigma-Aldrich

Octadecane

purum, ≥97.0% (GC)

  • CAS Number 593-45-3

  • Linear Formula CH3(CH2)16CH3

  • Molecular Weight 254.49

  •  Beilstein Registry Number 1740138

  •  EC Number 209-790-3

  •  MDL number MFCD00009007

Purchase

Properties

Related Categories Acyclic, Alkanes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
vapor density   8.8 (vs air)
vapor pressure   1 mmHg ( 119 °C)
InChI Key   RZJRJXONCZWCBN-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
autoignition temp.   455 °F
impurities   olefins, none detected
bp   317 °C(lit.)
mp   26-29 °C(lit.)
  26-29 °C
density   0.777 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
330.8 °F
Flash Point(C) 
166 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?