EMAIL THIS PAGE TO A FRIEND
74720 Sigma-Aldrich

1-Octadecanol

puriss., ≥99.0% (GC)

Synonym: Octadecyl alcohol, Stearyl alcohol

  • CAS Number 112-92-5

  • Linear Formula CH3(CH2)17OH

  • Molecular Weight 270.49

  •  Beilstein Registry Number 1362907

  •  EC Number 204-017-6

  •  MDL number MFCD00002823

Purchase

Properties

Related Categories Alcohols, Biochemicals and Reagents, Building Blocks, C11 to C30+, Chemical Synthesis,
vapor density   9.3 (vs air)
vapor pressure   <0.01 mmHg ( 38 °C)
InChI Key   GLDOVTGHNKAZLK-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
autoignition temp.   842 °F
expl. lim.   ~8 %
mp   56-59 °C(lit.)
  58-61 °C
solubility   methanol: soluble10 mg/mL, clear, colorless
density   0.812 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

WGK Germany 
nwg
RTECS 
RG2010000
Flash Point(F) 
383 °F
Flash Point(C) 
195 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?