EMAIL THIS PAGE TO A FRIEND
74725 Sigma-Aldrich

NZYM Broth

for microbiology

Purchase

Properties

Related Categories All Liquid Media/Broths, Escherichia coli & Coliforms, Media, Media by Microorganism, Media from A - Z,
grade   for microbiology
composition   magnesium sulfate heptahydrate, 2 g/L
  protein hydrolysate N-Z-amine, 10 g/L
  sodium chloride, 5 g/L
  yeast extract, 5 g/L
suitability   nonselective for Escherichia coli and Coliforms

Description

Application

For molecular biology.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?