EMAIL THIS PAGE TO A FRIEND
74745 Sigma-Aldrich

1-Octadecene

technical, ~85% (GC)

  • CAS Number 112-88-9

  • Linear Formula CH3(CH2)15CH=CH2

  • Molecular Weight 252.48

  •  Beilstein Registry Number 1761067

  •  EC Number 204-012-9

  •  MDL number MFCD00009003

  •  PubChem Substance ID 329765835

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
vapor density   0.71 (vs air)
InChI Key   CCCMONHAUSKTEQ-UHFFFAOYSA-N
grade   technical
assay   ~85% (GC)
autoignition temp.   482 °F
refractive index   n20/D 1.444(lit.)
  n20/D 1.445
bp   179 °C/15 mmHg(lit.)
  314.4 °C±5.0 °C/760 mmHg(lit.)
mp   14-16 °C(lit.)
density   0.789 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
300.2 °F
Flash Point(C) 
149 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

O806 1-Octadecene, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?