• USA Home
  • 74765 - Trimethyloctadecylammonium bromide

EMAIL THIS PAGE TO A FRIEND
74765 Sigma-Aldrich

Trimethyloctadecylammonium bromide

purum, ≥97.0% (AT)

Synonym: Octadecyltrimethylammonium bromide

  • CAS Number 1120-02-1

  • Linear Formula CH3(CH2)17N(Br)(CH3)3

  • Molecular Weight 392.50

  •  Beilstein Registry Number 3574090

  •  EC Number 214-294-5

  •  MDL number MFCD00043171

  •  PubChem Substance ID 329765847

Purchase

Properties

Related Categories Ammonium Salts, Biochemicals and Reagents, Cationic Detergents, Chemical Synthesis, Detergents,
description   cationic
InChI Key   SZEMGTQCPRNXEG-UHFFFAOYSA-M
grade   purum
assay   ≥97.0% (AT)
mp   ~250 °C (dec.)(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

359246 Trimethyloctadecylammonium bromide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?