• USA Home
  • 74835 - Dimethyl suberimidate dihydrochloride

EMAIL THIS PAGE TO A FRIEND
74835 Sigma-Aldrich

Dimethyl suberimidate dihydrochloride

≥98.0% (AT)

Synonym: DMS, Dimethyl ester octanediimidic acid dihydrochloride

Purchase

Properties

Related Categories Amidates/Imidates, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   ILKCDNKCNSNFMP-UHFFFAOYSA-N
assay   ≥98.0% (AT)
mp   213-214 °C(lit.)
  215-218 °C
Gene Information   rat ... Maoa(29253), Maob(25750)

Description

Other Notes

Cross-linking reagent for the modification of proteins via amidation; Lysine specific cross-linker.

Application

Reactant involved in:
• Chemical modification of tyrosinase for coupling to produce biocatalytic enzymatic flow-through detectors
• Cross-linker for synthesis of collagen-calcium phosphate composites

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?