• USA Home
  • 74853 - 4-(N,N-Dimethylsulfamoyl)-7-hydrazino-benzofurazan

EMAIL THIS PAGE TO A FRIEND
74853 Sigma-Aldrich

4-(N,N-Dimethylsulfamoyl)-7-hydrazino-benzofurazan

for fluorescence, ≥97.0% (CHN)

Synonym: 7-Hydrazino-N,N-dimethyl-4-benzofurazansulfonamide, DBD-H

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Benzofurazans, Derivatization Reagents, Derivatization Reagents HPLC,
grade   for fluorescence
InChI Key   VPDDJYGQJSTHTA-UHFFFAOYSA-N
assay   ≥97.0% (CHN)
fluorescence   λex 392 nm; λem 578 nm in 0.1 M phosphate pH 7.0
storage temp.   2-8°C

Description

Other Notes

Fluorogenic reagent for detecting aldehydes and ketones by HPLC; peroxyoxalate chemiluminescence detection

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?