EMAIL THIS PAGE TO A FRIEND
74863 Sigma-Aldrich

(S)-(+)-2-Octanol

for chiral derivatization

  • CAS Number 6169-06-8

  • Linear Formula CH3(CH2)5CH(OH)CH3

  • Molecular Weight 130.23

  •  Beilstein Registry Number 1719323

  •  EC Number 228-213-6

  •  MDL number MFCD00064283

  •  PubChem Substance ID 329765919

Purchase

Properties

Related Categories Alcohols, Analytical Reagents, Analytical/Chromatography, Asymmetric Synthesis, Chemical Synthesis,
grade   for chiral derivatization
InChI Key   SJWFXCIHNDVPSH-QMMMGPOBSA-N
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D +10.0±0.5°, neat
optical purity   enantiomeric ratio: ≥99.5:0.5 (GC)
impurities   ≤0.5% water
refractive index   n20/D 1.426(lit.)
  n20/D 1.426
bp   175 °C(lit.)
density   0.820 g/mL at 20 °C
  0.822 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

147982 (S)-(+)-2-Octanol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?